missing translation for 'onlineSavingsMsg'
Learn More
Sulfate Standard Solution, 1000μg/mL, Ion HIQU, Chem-Lab Product Code.: 11959122

Sulfate Standard Solution, 1000μg/mL, Ion HIQU, Chem-Lab

Product Code. p-8014416
100 mL, 100mL
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11959122

Brand: Chem Lab CL01.1971.0100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a reference and calibration standard in laboratory applications. Sulfate Standard Solution, 1000μg/mL, Ion HIQU, Chem-Lab is prepared from 1.479g Na2SO4 per liter of water to contain a precise concentration of SO42- ions for use with ion-selective electrodes.

This sulfate standard solution contains a precise concentration of 1000μg/mL SO42− (±7μg at 20°C). It is used to verify the accuracy of sulfate determinations and to calibrate equipment used in those determinations.

Chemical Identifiers

CAS 7757-82-6, 7732-18-5
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym Sodium Sulfate Solution, Sulfate of Soda
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Description Chloride: <0,02mg/L
Iodide: <0.01mg/L
Nitrite: <0.01mg/L
Nitrate: <0.01mg/L
Phosphate: <0.02mg/L
Density 1.00g/mL
Concentration 1000 μg/mL of SO42−
IUPAC Name disodium;sulfate
Quantity 100 mL
Formula Weight 142.04g/mol
Concentration or Composition (by Analyte or Components) 1.479g Na2SO4 per 1L H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfate Standard Solution, 1000μg/mL, Ion HIQU, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.