Få mere at vide
Sulfanitran-d4 (Major) (>85%), TRC
Artikelnummer. 30413230
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413230 25 mg
30427883 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413230 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68904225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1794753-46-0
Molekylær formel C14H9D4N3O5S
Molekylvægt (g/mol) 339.36
Synonym 4'-[(p-Nitrophenyl)sulfamoyl]acetanilide-d4, N4-Acetyl-4'-nitrosulfanilanilide-d4, N4-Acetyl-N1-(4-nitrophenyl)sulfanilamide-d4, N4-Acetyl-N1-(p-nitrophenyl)sulfanilamide-d4, NSC 217299-d4, NSC 77120-d4
SMIL O=S(C1=C([2H])C([2H])=C(NC(C)=O)C([2H])=C1[2H])(NC2=CC=C([N+]([O-])=O)C=C2)=O
Tekniske data
Kemisk navn eller materiale Sulfanitran-d4
Mængde 25 mg
InChI formel InChI=1S/C14H13N3O5S/c1-10(18)15-11-4-8-14(9-5-11)23(21,22)16-12-2-6-13(7-3-12)17(19)20/h2-9,16H,1H3,(H,15,18)/i4D,5D,8D,9D
Formel vægt 339.0827