Få mere at vide
Sulfametrole, TRC
Artikelnummer. 30138071
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138071 100 mg
30006930 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138071 Leverandør Toronto Research Chemicals Leverandørnr. TRCS699025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 32909-92-5
Molekylær formel C9 H10 N4 O3 S2
Molekylvægt (g/mol) 286.33
Synonym Benzenesulfonamide, 4-amino-N-(4-methoxy-1,2,5-thiadiazol-3-yl)-, Sulfanilamide, N1-(4-methoxy-1,2,5-thiadiazol-3-yl)- (7CI,8CI), 1,2,5-Thiadiazole, benzenesulfonamide deriv., 4-Amino-N-(4-methoxy-1,2,5-thiadiazol-3-yl)benzenesulfonamide, Sulfametrole
IUPAC navn 4-amino-N-(4-methoxy-1,2,5-thiadiazol-3-yl)benzenesulfonamide
SMIL COc1nsnc1NS(=O)(=O)c2ccc(N)cc2
Tekniske data
Kemisk navn eller materiale Sulfametrole
Mængde 100 mg
InChI formel InChI=1S/C9H10N4O3S2/c1-16-9-8(11-17-12-9)13-18(14,15)7-4-2-6(10)3-5-7/h2-5H,10H2,1H3,(H,11,13)
Formel vægt 286.0194