Få mere at vide
Sulfadiazine Sodium Salt, TRC
Artikelnummer. 17934205
Skift visning
Mængde:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17934205 50 g
17964425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17934205 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69905850G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 547-32-0
Molekylær formel C10H9N4NaO2S
Molekylvægt (g/mol) 272.26
Synonym 2-Sulfanilamidopyrimidine Sodium Salt, Monosodium 2-Sulfanilamidopyrimidine, Sodium 2-Sulfanilamidopyrimidine, Sodium Sulfadiazine, Sodium Sulfapyrimidine, Soluble Sulfadiazine, Soludiazine, Sulfadiazine Sodium, Sulfadiazine Sodium Salt, Sulfapyrimidine Sodium, Suthogen, N1-2-Pyrimidinyl-sulfanilamide Monosodium Salt, (N1-2-Pyrimidinylsulfanilamido)-sodium, 4-Amino-N-2-pyrimidinyl-benzenesulfonamide Monosodium Salt
IUPAC navn sodium;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide
SMIL [Na+].Nc1ccc(cc1)S(=O)(=O)[N-]c2ncccn2
Tekniske data
Kemisk navn eller materiale Sulfadiazine Sodium Salt
Mængde 50 g
InChI formel InChI=1S/C10H9N4O2S.Na/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10;/h1-7H,11H2;/q-1;+1
Formel vægt 272.0344