missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Sulfacetamide Sodium, MedChemExpress
Beskrivelse
Sulfacetamide Sodium is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sulfacetamide Sodium |
| CAS | 127-56-0 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C8H9N2NaO3S |
| Mængde | 5 g |
| Opløselighedsinformation | H2O : ≥ 50 mg/mL (211.67 mM) ∣DMSO : 33.33 mg/mL (141.10 mM; Need ultrasonic) |
| SMIL | O=S(C1=CC=C(N)C=C1)(N([Na])C(C)=O)=O |
| Molekylvægt (g/mol) | 236.22 |
| Formel vægt | 236.22 |
| Vis mere |
Research purposes only
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?