Learn More
Kartell™ Suction Tube
Kartell™ Suction Tube is an accessory for a Dosipet or Technotrate and is used to obtain quantities of a liquid.
Brand: Kartell™ 8301400
Additional Details : Weight : 1.00000kg
Description
2.5-10mL for Dosipet dispenserSpecifications
Suction Tube | |
2.5 to 10 mL | |
Dosipet |