missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Suction Tube

Kartell™ Suction Tube is an accessory for a Dosipet or Technotrate and is used to obtain quantities of a liquid.

126.00 DKK - 198.00 DKK

Specifications

Description Suction Tube
For Use With (Equipment) Dosipet
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Volume (Metric) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Volume (Metric) Price Quantity & Availability  
13181094
View Documents Promotion Details
Kartell™
8301400
Dosipet 2.5 to 10 mL
126.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
13191094
View Documents Promotion Details
Kartell™
8301500
Dosipet 25 to 100 mL
198.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

  • Accessory for Dosipet and Technotrate
Specifications

Specifications

Suction Tube
Dosipet
Videos
SDS
Documents

Documents

Product Certifications