Få mere at vide
Sucrose Hexasulfate, Potassium Salt, Technical Grade, TRC
Artikelnummer. 30134521
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134521 250 mg
17959904 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134521 Leverandør Toronto Research Chemicals Leverandørnr. TRCS698350250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 386229-70-5
Molekylær formel C12 H16 O29 S6 . 6 K
Molekylvægt (g/mol) 1051.22
Synonym Sucrose Hexasulfate, Potassium Salt
IUPAC navn hexapotassium;[(2R,3R,4S,5R,6R)-2-[(2S,3S,4R,5R)-3,4-disulfonatooxy-2,5-bis(sulfonatooxymethyl)oxolan-2-yl]oxy-5-hydroxy-6-(hydroxymethyl)-3-sulfonatooxyoxan-4-yl] sulfate
SMIL [K+].[K+].[K+].[K+].[K+].[K+].OC[C@H]1O[C@H](O[C@]2(COS(=O)(=O)[O-])O[C@H](COS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@@H]2OS(=O)(=O)[O-])[C@H](OS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@@H]1O
Tekniske data
Kemisk navn eller materiale Sucrose Hexasulfate, Potassium Salt, Technical Grade
Mængde 250 mg
InChI formel InChI=1S/C12H22O29S6.6K/c13-1-4-6(14)8(39-45(24,25)26)9(40-46(27,28)29)11(35-4)37-12(3-34-43(18,19)20)10(41-47(30,31)32)7(38-44(21,22)23)5(36-12)2-33-42(15,16)17;;;;;;/h4-11,13-14H,1-3H2,(H,15,16,17)(H,18,19,20)(H,21,22,23)(H,24,25,26)(H,27,28,29)(H,30,31,32);;;;;;/q;6*+1/p-6/t4-,5-,6-,7-,8+,9-,10+,11-,12+;;;;;;/m1....../s1
Formel vægt 1049.5924