missing translation for 'onlineSavingsMsg'
Learn More

SU6656, MedChemExpress

Product Code. 16444487
Encompass_Preferred
Change view
Click to view available options
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16444487 50 mg
16421068 5 mg
16408618 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444487 Supplier Medchem Express Supplier No. HYB0789

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SU6656 is a Src family kinases inhibitor with IC50s of 280, 20, 130, 170 nM for Src, Yes, Lyn, and Fyn, respectively. SU6656 inhibits FAK phosphorylation at Y576/577, Y925, Y861 sites. SU6656 also inhibits p-AKT.

SU6656 is a Src family kinases inhibitor with IC50s of 280, 20, 130, 170 nM for Src, Yes, Lyn, and Fyn, respectively. SU6656 inhibits FAK phosphorylation at Y576/577, Y925, Y861 sites. SU6656 also inhibits p-AKT.

Specifications

Kemisk navn eller materiale SU6656
CAS 330161-87-0
Farve Orange
Renhedsgrad noter Research
Molekylær formel C19H21N3O3S
Mængde 50 mg
Opløselighedsinformation DMSO : 20 mg/mL (53.84 mM; Need ultrasonic)
SMIL O=S(C1=CC2=C(NC(/C2=C\C(N3)=CC4=C3CCCC4)=O)C=C1)(N(C)C)=O
Molekylvægt (g/mol) 371.45
Formel vægt 371.45
Procent renhed 96.87%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.