Learn More
IBA Lifesciences Strep-Tactin™ PE or APC
Shop All IBA Lifesciences Products
Click to view available options
Label or Dye:
APC Conjugate
PE Conjugate
Unit Size:
50µL
Description
- Strep-Tactin™ conjugated to either R-Phycoerythrin (PE) or Allophycocyanin (APC) is suitable for staining or isolation of antigen-specific cytotoxic T cells (MHC I Streptamer™) or surface marker-specific cells (Fab-TACS™/Nano-TACS™) based on IBAs Strep-tag™ technology.
- To target specific cells, MHC I complexes, low affinity Fab fragments or nanobodies fused to a Twin-Strep-tag™ (MHC I-Streps, Fab-Streps or Nano-Streps) are required in addition to fluorescent Strep-Tactin™.
- Strep-Tactin™ multimerizes MHC I-Streps or Fab-/Nano-Streps, ensuring stable binding to cells.
- The resulting fluorescent complexes can be used to stain cells that are specific for the respective peptide-loaded MHC I molecule or for a selected surface marker.
- The addition of biotin reverses the multimerization and causes the dissociation of MHC I-Streps or Fab-/Nano-Streps from the cell surface.
- The complete removal of all labeling reagents preserves the cells’ full biological activity.
Greener Choice Claims
- Environmental benefits include:
- Non-Toxic
Specifications
Specifications
Kit Contents | 50 tests a 1 to 2 x106 cells |
Quantity | 50 Tests |
Label or Dye | APC Conjugate |
Documents
Product Information
Manuals
Manual PR22 MHC I Streptamer 6-5010-001
File size: 1.2 MB
Methods & Protocols
Short Protocol PR38 MHC-I Staining 6-5010-001
File size: 1.1 MB
For Research Use Only.
Product Content Correction
Product Title
IBA Lifesciences Strep-Tactin™ PE or APC >
Spot an opportunity for improvement?Share a Content Correction