Learn More
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10326231
![]() |
Thermo Scientific Nalgene
715311-0250 |
Cover for port on dessicator, polycarbonate |
3335.00 DKK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Get the Stopcock Assembly and Cover replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator, Cat #5311-0250. The Nalgene™ Transparent Polycarbonate Classic Design Desiccator maintains an airtight seal without the need of vacuum grease and can be used under full-vacuum and non-vacuum conditions, and is an excellent alternative to glass.
DescripciĂłn:
- Replacement stopcock for Nalgene desiccator item number 5311-0250
- Replacement cover for Nalgene desiccator item number 5311-0250
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.