Få mere at vide
Stigmastanol, MedChemExpress
Artikelnummer. 16497517
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16497517 5 mg
16484778 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16497517 Leverandør Medchem Express Leverandørnr. HY113494
MedChemExpress Stigmastanol is the 6-amino derivative isolated from Hypericum riparium. Hypericum riparium A. Chev. is a Cameroonian medicinal plant belonging to the family Guttiferae.
Stigmastanol is the 6-amino derivative isolated from Hypericum riparium. Hypericum riparium A. Chev. is a Cameroonian medicinal plant belonging to the family Guttiferae.
Tekniske data
Kemisk navn eller materiale Stigmastanol
CAS 83-45-4
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C29H52O
Mængde 5 mg
Opløselighedsinformation Ethanol : 5 mg/mL (12.00 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.40 mM; ultrasonic and warming and heat to 80°C)
SMIL CC[C@H](CC[C@@H](C)[C@]1([H])[C@]2(C)[C@](CC1)([H])[C@]3([H])CC[C@@]4([H])C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC2)C(C)C
Molekylvægt (g/mol) 416.72
Formel vægt 416.72
Procent renhed 93.0%
Grad Research
Fysisk form Solid