Learn More
Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes
Conduct applications with the chemical resistance of Pyrex™ borosilicate glass using Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes, in easy to handle shelf packs.
3915.00 DKK
Product Code | Brand | Description | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Price | Quantity & Availability | ||||
10663961
|
Thermo Scientific™
7079-2N |
Pipettes, unplugged | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Conduct applications with the chemical resistance of Pyrex™ borosilicate glass using Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes, in easy to handle shelf packs.
Features:- Calibrated for delivery with the last drop expelled by blowing
- Available either plugged or unplugged
- Easy to read graduations
Ordering Information:
Product is not available in all geographic regions.
Specifications
Glass | |
Yes | |
290 mm |