Learn More
Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes
Conduct applications with the chemical resistance of Pyrex™ borosilicate glass using Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes, in easy to handle shelf packs.
Brand:  Thermo Scientific™ 7079-2N
Additional Details : Weight : 7.80000kg
 View more versions of this product
Product Code. 10663961
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Conduct applications with the chemical resistance of Pyrex™ borosilicate glass using Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes, in easy to handle shelf packs.
  • Bulk packed in easy-to-handle shelf packs
  • Calibrated for delivery with the last drop expelled by blowing
  • Available either plugged or unplugged
  • Easy-to-read graduations
Ordering Alerts:
Not available in all geographic regions
Specifications
Specifications
Pipettes, unplugged
Non-sterile
-0.2 mL
2 mL
Glass
Yes
35/Pk., 700/Cs.
290 mm