missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1043.29 DKK - 2973.37 DKK
Specifications
Antigen | STEAP1 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16859714
|
Proteintech
20199-1-AP-20UL |
20 μL |
1043.29 DKK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16849714
|
Proteintech
20199-1-AP-150UL |
150 μL |
2973.37 DKK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
STEAP1 is predomitly expressed in prostate tissue, and is found to be upregulated in multiple cancer cell lines. The gene product is predicted to be a six-transmembrane protein, and was shown to be a cell surface antigen significantly expressed at cell-cell junctions.Specifications
STEAP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UHE8 | |
26872 | |
STEAP1 Fusion Protein Ag14134 | |
Primary | |
-20°C | |
STEAP1 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Metalloreductase STEAP1, PRSS24, STEAP, STEAP1 | |
STEAP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |