missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20199-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
STEAP1 is predomitly expressed in prostate tissue, and is found to be upregulated in multiple cancer cell lines. The gene product is predicted to be a six-transmembrane protein, and was shown to be a cell surface antigen significantly expressed at cell-cell junctions.Specifications
STEAP1 | |
Polyclonal | |
Unconjugated | |
STEAP1 | |
Metalloreductase STEAP1, PRSS24, STEAP, STEAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26872 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UHE8 | |
STEAP1 | |
STEAP1 Fusion Protein Ag14134 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |