Få mere at vide
IBA Lifesciences StarGate pLSG-IBA Vectors for Baculo
GREENER_CHOICE
Artikelnummer. 15643828 Shop alle IBA Lifesciences produkter
Vektor:
pLSG-IBA 102
pLSG-IBA 103
pLSG-IBA 104
pLSG-IBA 105
pLSG-IBA 123
pLSG-IBA 142
pLSG-IBA 143
pLSG-IBA 144
pLSG-IBA 145
pLSG-IBA 162
pLSG-IBA 164
pLSG-IBA 167
pLSG-IBA 168
pLSG-IBA 23
pLSG-IBA 25
pLSG-IBA 3
pLSG-IBA 33
pLSG-IBA 35
pLSG-IBA 43
pLSG-IBA 45
pLSG-IBA 5
pLSG-IBA 62
pLSG-IBA 63
pLSG-IBA 64
pLSG-IBA 65
pLSG-IBA wt1
pLSG-IBA wt2
Pakningsstørrelse:
5µg
Artikelnummer. 15643828
Brand: IBA Lifesciences 54803001
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Transfer vectors that allow for high-level expression in insect cells by introducing the gene of interest into the Polyhedrin gene locus of AcMNPV DNA by homologous recombination.
pLSG-IBA Baculo virus (insect cells) expression vectors
The pLSG-IBAwt1 vector carries the Polyhedrin promoter for high-level expression in insect cells.
  • Co-transfection with BacPAK6 linearized AcMNPV DNA (Clontech) or circular flashBAC modified AcMNPV DNA (Oxford Expression Technologies) allows the generation of recombinant baculovirus at very high efficiency through reconstitution of an essential gene (ORF 1629) and elimination of wild type virus to great extent.
  • Cloning and expression of recombinant proteins in insect cells
  • Polyhedrin promoter drives high-level expression of the insert
  • Ampicillin resistance and ColE1 origin of replication (pUC) support propagation in E. coli
Please note that cloning into pLSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pLSG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.
Tekniske data
Format Væske
Til brug med (udstyr) StarGate™ System, Insect Host Cells
Inducerende middel BacPAK6 linearized A cmNPV DNA (Clontech) or circular flashBAC modified A cmNPV DNA (Oxford Expression Technologies)
Inkluderer pUC ori, Ampicillin resistance
pH 8
Promotor Polyhedrin
Produkttype Acceptor Vector for bacula (insect cell) expression
Opbevaringsbuffer 20μ L TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5μ g
Indhold og opbevaring 2 °C til °8 C ved hyppig °brug, -20 C til langtidsopbevaring
Protein tag Strep-tag for C-terminal
Koncentration 250 ng/μ L
Mængde 5μ g
Vektor pLSG-IBA 3
Farve Klar
Til brug med (applikation) Kloning af gener af interesse til brug ved funktionel ekspression, oprensning, påvisning, immobilisering eller adskillelse af rekombinante proteiner