Learn More
Karl Hecht™ Assistent™ Tubing Clamps
Provides for safe, secure and easy connection between tubing in the laboratory. Karl Hecht™ Assistent™ Tubing Clamps are manufactured from stainless steel.
Brand: Karl Hecht™ 41586021
Additional Details : Weight : 0.02000kg
Description
- With thumb screw
- Available for tubing of various diameters
Spécification
Tubing Clamp | |
Stainless Steel | |
For 21mm tubing I.D. | |
1 Each |