Learn More
Sartorius™ Stainless Steel OIML Class F2 Reference Line Weight
Stainless Steel OIML Class F2 Reference Line Weight without DAkkS Calibration Certificate.
Brand: Sartorius™ YCW314-00
434.23 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Without DAkkS Calibration Certificate
Specifications
Stainless Steel | |
Plastic | |
1 g |
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F2 Reference Line Weight > Plastic; Mass: 1 g
Spot an opportunity for improvement?Share a Content Correction