Learn More
Sartorius™ Stainless Steel OIML Class F2 Calibration Weight Without DAkkS Calibration Certificate
Highly polished steel calibration weights from 100g to 10kg
Brand:  Sartorius™ YCW5148-00
820.71 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10591541
  • 846.00 DKK / Each
Only null left
Estimated Shipment: 11-07-2025
Description
To OIML Class E2, F1 and F2 specification which includes DKD (Deutscher Kalibrierdienst accredited by the PTB) certificate (UKAS Equivalent).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plastic
100 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F2 Calibration Weight Without DAkkS Calibration Certificate > Mass: 100g