Learn More
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW312-02
1551.88 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10328872
  • 1595.00 DKK / Each
Only null left
Estimated Shipment: 12-08-2025
Specifications
Wood
1 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate > Mass: 1g