Learn More
Sampling Systems™ Stainless Steel Mini-Drums without Toggle Clamps
These Mini-Drums without Toggle Clamps are excellent multi-purpose containers for a wide variety of applications in the laboratory.
Brand: Sampling Systems™ A434-8
Additional Details : Weight : 5.95820kg
Description
- Manufactured from durable 304 grade stainless steel
- Crevice-free interior
- Neat recessed fitting lid
- Complete with a swing carrying handle
- Flat bottom with chime ring to give added protection
- Available in a wide range of capacities to fit your specific needs
Spécification
Mini Drum | |
Recessed Lid | |
Multi-purpose Storage | |
Swing Handle | |
200 mm |
Yes | |
8 L | |
Stainless Steel | |
285 mm |