Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand:  Hamilton™ 7751-12
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11513264
  • 2660.00 DKK / Pack of 6
Estimated Shipment: 02-05-2024
to see stock.
Description
Description
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Specifications
Autoclavable
21 G
0.82 mm
6/Pk.
5 or AS
Standard Tapered
Nickel-plated Brass (Hub), Stainless Steel
0.51 mm
Luer Lock Custom Needle
9.52 to 304 mm
TLL Syringes and LT or TLL Connectors