Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
2660.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Nickel-plated Brass (Hub), Stainless Steel |
Product Type | Luer Lock Custom Needle |
Quantity | 6/Pk. |
Needle Point Style | 5 or AS |
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
11553264
|
Hamilton™
7751-16 |
25 G |
2660.00 DKK
Pack of 6 |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11513264
|
Hamilton™
7751-12 |
21 G |
2660.00 DKK
Pack of 6 |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11503264
|
Hamilton™
7751-11 |
20 G |
2660.00 DKK
Pack of 6 |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Autoclavable | |
Luer Lock Custom Needle | |
5 or AS | |
Standard Tapered |
Nickel-plated Brass (Hub), Stainless Steel | |
6/Pk. | |
TLL Syringes and LT or TLL Connectors |