Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand:  Ohaus™ 30517705
653.57 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16672292
  • 673.00 DKK / Each
Only null left
Estimated Shipment: 29-07-2025
Description
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Certificate
2 mg
Protective Case
Stainless Steel (316)
0.003 mg
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire > 2 mg