Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand: Ohaus™ 30517960
653.57 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
No Certificate | |
100 mg | |
Protective Case |
Stainless Steel (316) | |
0.005 mg |
Product Content Correction
Product Title
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire > 100 mg
Spot an opportunity for improvement?Share a Content Correction