Learn More
3M™ Speedglas™ Outer Protection Plate 9000
3M™ Speedglas™ Outer Protection Plate 9000 for use with 3M™ Speedglas™ Welding Helmet Series 9000.
480.00 DKK - 4325.00 DKK
Specifications
Certifications/Compliance | EN 166, EN166:BT |
---|---|
For Use With (Equipment) | Speedglas™ Welding Helmet Series 9000 |
Lens Tint | Clear |
Product Type | Outer Protection Plate |
Product Code | Brand | Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Style | Price | Quantity & Availability | |||||
15729637
|
3M™
7000000206 |
Heat Resistant |
525.00 DKK
Pack of 10 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
15780048
|
3M™
7100016860 |
Standard |
4325.00 DKK
Pack of 10 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
15799627
|
3M™
7000000183 |
Scratch Resistant |
480.00 DKK
Pack of 10 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
Description
- Replacement spare part to the original protection plate if any pits or scratches occur
- For maximum safety and comfort, make it a habit to check your equipment regularly
- Regularly clean with a soft cloth and replace if any pits or scratches occur
Specifications
EN 166, EN166:BT | |
Speedglas™ Welding Helmet Series 9000 | |
Clear | |
Outer Protection Plate |