Få mere at vide
Sorafenib N-Oxide, TRC
Artikelnummer. 30133777
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133777 1 mg
17972655 25 mg
17952195 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133777 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6768701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 583840-03-3
Molekylær formel C21 H16 Cl F3 N4 O4
Molekylvægt (g/mol) 480.82
Synonym 2-Pyridinecarboxamide, 4-[4-[[[[4-chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-, 1-oxide, 4-[4-[[4-Chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]-N-methyl-1-oxido-pyridin-1-ium-2-carboxamide, Sorafenib N-Oxide
IUPAC navn 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]-N-methyl-1-oxidopyridin-1-ium-2-carboxamide
SMIL CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)cc2)cc[n+]1[O-]
Tekniske data
Kemisk navn eller materiale Sorafenib N-Oxide
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C21H16ClF3N4O4/c1-26-19(30)18-11-15(8-9-29(18)32)33-14-5-2-12(3-6-14)27-20(31)28-13-4-7-17(22)16(10-13)21(23,24)25/h2-11H,1H3,(H,26,30)(H2,27,28,31)
Formel vægt 480.0812
Procent renhed >95%