Learn More
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For Vo49
Memmert™ AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand: Memmert™ VO49FDAQ1
This item is not returnable.
View return policy
Description
- Software conforming to FDA AtmoCONTROL
- Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
- Base licence for the control of one unit
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA) | |
Models U, UF, TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 106 | |
Software |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Memmert AtmoCONTROL (EN)
-
Memmert ControlCOCKPIT (EN)
Product Content Correction
Product Title
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For Vo49 >
Spot an opportunity for improvement?Share a Content Correction