Få mere at vide
Sodium Propionate-13C3, TRC
Artikelnummer. 30136044
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136044 1 mg
30138310 2 mg
30009103 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136044 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6732901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 152571-51-2
Molekylær formel 13C3H5NaO2
Molekylvægt (g/mol) 99.04
Synonym Propanoic-13C3 acid, sodium salt (9CI), Sodium propionate-13C3, Sodium propanoate-13C3, Propionic acid-1,2,3-13C3 sodium salt
SMIL [13CH3][13CH2][13C](O[Na])=O
Tekniske data
Kemisk navn eller materiale Sodium Propionate-13C3
Mængde 1 mg
InChI formel InChI=1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1/i1+1,2+1,3+1;
Formel vægt 99.0288