missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hypochlorite Solution, 47/50%, Chem-Lab Product Code.: 11909012
compliance-icons

Sodium Hypochlorite Solution, 47/50%, Chem-Lab

Product Code. p-8014301
1 L, 1 L
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
This item is not returnable. View return policy

Product Code. 11909012

Brand: Chem Lab CL00.1410.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a disinfectant and bleaching agent. Sodium Hypochlorite Solution, 47/50%, Chem-Lab is a chemical compound also known as bleach. It is a common laboratory reagent that reacts with many acids and is a strong oxidizer.

Store in a cool environment, at 2–15°C.

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solutionShow MoreShow Less
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Yellow
Boiling Point 100°C
Melting Point 0°C
Composition Available chlorine: ± 13%
Density 1.22g/mL
Solubility Information Water Soluble
Formula Weight 74.44g/mol
Percent Purity 47/50% (NaClO)
Odor Chlorine
pH >13
Quantity 1 L
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hypochlorite 47/50% solution p.
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hypochlorite Solution, 47/50%, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.