Learn More
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ Product Code.: 10438410
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Product Code. 10438410 Shop All Fisher Chemical Products
5 L, Plastic container
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Product Code. 10438410
Brand: Fisher Chemical J/7630/21
Request Bulk or Custom Format
Sodium Hydroxide Solution , NIST standard solution ready to use, for volumetric analysis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, causticShow MoreShow Less
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
Specifications
pH 12
Color Colorless
Boiling Point 100°C
Melting Point 0°C
Quantity 5 L
Packaging Plastic container
Vapor Density > 1.0
Physical Form Liquid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2539414 Certificate TypeCertificate of Analysis Date09/06/2025 Catalog NumberJ/7630/24,J/7630C/90,J/7630/15,J/7630/27,J/7630/17,J/7630C/05,J/7630/21
Lot Number2537014 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberJ/7630/24,J/7630C/90,J/7630/15,J/7630/27,J/7630/17,J/7630C/05,J/7630/21
Lot Number2533959 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberJ/7630/24,J/7630C/90,J/7630/15,J/7630/27,J/7630/17,J/7630C/05,J/7630/21
Lot Number2531391 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberJ/7630/24,J/7630C/90,J/7630/15,J/7630/27,J/7630/17,J/7630C/05,J/7630/21
compliance-icons
Product Identifier
  • Sodium hydroxide, solution 0.5M
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P390-Absorb spillage to prevent material damage.
Product Content Correction
Product Title
Sodium Hydroxide Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ >