missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hexametaphosphate, SLR, Powder, Mainly (NaPO3)6, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Description
Na6O18P6, CAS Number-10124-56-8, 500g, White, 611.7708g/mol, 640 deg.C, 136045, Plastic powder jar, 6.0 to 7.7, Solid

Specifications
Specifications
Melting Point | 640°C |
Color | White |
pH | 6.0 to 7.7 |
Physical Form | Powder |
Quantity | 500 g |
Formula Weight | 611.7708g/mol |
Packaging | Plastic powder jar |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2539763 | Certificate of Analysis | 09/06/2025 | S/4160/53,S/4160/60 |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hexametaphosphate, SLR, Powder, Mainly (NaPO3)6, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction