Få mere at vide
Sodium Dibromomethanesulfonate, TRC
Artikelnummer. 17972485
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17972485 50 mg
17911475 10 mg
17964695 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17972485 Leverandør Toronto Research Chemicals Leverandørnr. TRCS62411050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 859073-88-4
Molekylær formel CHBr2NaO3S
Molekylvægt (g/mol) 275.88
Synonym 1,1-Dibromomethanesulfonic Acid Sodium Salt
IUPAC navn sodium;dibromomethanesulfonate
SMIL [Na+].[O-]S(=O)(=O)C(Br)Br
Tekniske data
Kemisk navn eller materiale Sodium Dibromomethanesulfonate
Mængde 50 mg
InChI formel InChI=1S/CH2Br2O3S.Na/c2-1(3)7(4,5)6;/h1H,(H,4,5,6);/q;+1/p-1
Formel vægt 273.7911