Få mere at vide
Sodium Butyrate-13C4, TRC
Artikelnummer. 30012853
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012853 10 mg
30006009 2 mg
30005942 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012853 Leverandør Toronto Research Chemicals Leverandørnr. TRCS61517810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 312623-84-0
Molekylær formel 13C4H7NaO2
Molekylvægt (g/mol) 114.06
Synonym Butanoic-13C4 acid, sodium salt (9CI), Sodium butanoate-1,2,3,4-13C4, Butanoic-1,2,3,4-13C4 acid sodium salt, Sodium butanoate-13C4
SMIL [13CH3][13CH2][13CH2][13C](O[Na])=O
Tekniske data
Kemisk navn eller materiale Sodium Butyrate-13C4
Mængde 10 mg
InChI formel InChI=1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1/i1+1,2+1,3+1,4+1;
Formel vægt 114.0478