Få mere at vide
SNRNP25 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19890-1-AP-150UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The SNRNP25 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SNRNP25. This antibody recognizes human, mouse antigen. The SNRNP25 antibody has been validated for the following applications: IHC, ELISA analysis.Tekniske data
| SNRNP25 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 79622 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Immunohistochemistry, ELISA | |
| 300 μg/mL | |
| C16orf33; FLJ22940 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Polyclonal Antibody | |
| IgG |