Learn More
Chemsplash Snood Cap Peak Disposable
Snood Caps are a polypropylene heavyweight material that is also available in cotton or nylon.
2175.00 DKK
Specifications
Product Type | Snood Cap |
---|---|
Size | One Size |
Material | Polypropylene |
Closure Type | Ties |
Disposable | Single-use |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
30058382
![]() |
Chemsplash
2528.300.999 |
Blue |
2175.00 DKK
Pack of 1000 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
30058431
![]() |
Chemsplash
2528.400.999 |
White |
2175.00 DKK
Pack of 1000 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
Description
Snood Caps
- Heavyweight Polypropylene
- PFAS Free
Description: Snood Caps are made from a heavyweight polypropylene material, and they are also available in cotton or nylon.
Specifications
Snood Cap | |
Polypropylene | |
Single-use | |
Non-sterile |
One Size | |
Ties | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Chemsplash Snood Cap Peak Disposable