missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SNAP47 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56894
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SNAP47 Polyclonal specifically detects SNAP47 in Human samples. It is validated for Western Blot.
Tekniske data
| SNAP47 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C1orf142, Epididymis luminal protein 170, FLJ12517, HEL170, SNAP-47chromosome 1 open reading frame 142, SVAP1DKFZp686M10160, Synaptosomal-associated 47 kDa protein, synaptosomal-associated protein 47, synaptosomal-associated protein, 47kDa | |
| Rabbit | |
| 52.5 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q5SQN1 | |
| SNAP47 | |
| Synthetic peptides corresponding to C1ORF142 The peptide sequence was selected from the middle region of C1ORF142 (NP_444280). Peptide sequence TALHLQTSLPALSEADTQELTQILRRMKGLALEAESELERQDEALDGVAA. | |
| Affinity purified | |
| RUO | |
| 116841 | |
| Human, Mouse, Rat, Pig, Bovine, Equine, Rabbit, Yeast, Zebrafish | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion