missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SNAP29 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
3675.00 DKK
Tekniske data
| Antigen | SNAP29 |
|---|---|
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Værtsarter | Rabbit |
Beskrivelse
SNAP29 Polyclonal specifically detects SNAP29 in Rat samples. It is validated for Western Blot.Tekniske data
| SNAP29 | |
| Polyclonal | |
| Rabbit | |
| NP_446262 | |
| 9342 | |
| The immunogen for this antibody is Snap29. Peptide sequence NSIKSVFGGFINYFKSKPVEPPPEQNGSIVPQPSSRLKEAINTSKDQESK. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CEDNIKSoluble 29 kDa NSF attachment protein, SNAP-29FLJ21051, synaptosomal-associated protein 29, synaptosomal-associated protein, 29kD, synaptosomal-associated protein, 29kDa, Vesicle-membrane fusion protein SNAP-29 | |
| SNAP29 | |
| IgG | |
| 29 kDa |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel