Få mere at vide
SLC22A10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91568
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SLC22A10 Polyclonal specifically detects SLC22A10 in Human samples. It is validated for Western Blot.
Tekniske data
| SLC22A10 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| member 10, solute carrier family 22, member 10 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 387775 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_001034841 | |
| SLC22A10 | |
| Synthetic peptide directed towards the middle region of human SLC22A10. Peptide sequence QTLRVALACLGIGCSAATFSSVAVHFIELIPTVLRARASGIDLTASRIGA. | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |