Få mere at vide
Bollé Safety Slam sikkerhedsbriller
Shop alle Bollé Safety produkter
Linsefarve:
Skygge 1.7
Skygge 3
Skygge 5
Yellow
Linsebelægning:
Anti-fog, Impact-resistant, Scratch-resistant
Slagfast, Ridsefast
Pakningsstørrelse:
Hver
Beskrivelse
- Stylish.
- Easy to wear.
- Protection at all times.
- Unbeatable comfort.
- EN 166: Guaranteed minimum resistance of the protective eyewear against everyday risks including dropping the protective eyewear onto the ground, ageing by light exposure, exposure to heat or corrosion etc.
- EN 170, Risks relating to radiation - UV protection: The protection filters ultraviolet radiation. This type of protection is particularly recommended during activities where the source of ultraviolet radiation is predominant, such as mercury vapor lamps, actinic or germicides. Levels of application: 2-1.2, 2C-1.2 Lens color: Clear, blue, yellow or green. Adding the code 'C' indicates good color recognition by the holder.
- Mechanical risk F - Mechanical risk - Low energy impact:& Low energy impact, resists a 6 mm, 0.86 g ball at 45 m/s.
- Mechanical risk T - High speed particles at extreme temperatures: The letter T, immediately after the mechanical strength symbol, authorizes use for high-speed particles at extreme temperatures.
- Optical class: Class 1, continuous work, worn permanently.
- CE, European standard: Frame marked to indicate meeting CE standard.
Tekniske data
Tekniske data
| Linsebelægning | Anti-fog, Impact-resistant, Scratch-resistant |
| Oversigt | Model SLAPSJ |
| Certificeringer/overholdelse (Europa) | EN166, EN170/2C-1.2, Mechanical risk F/T, Optical class 1, CE |
| Linsefarve | Yellow |
| Produkttype | Safety glasses |
| Til brug med (applikation) | Indoor/cloudy sky, protection, continuous wear |
| nr. pr. sag | 200/Cs. |
| nr. pr. pakke | 1/bag, 10/Pk. |
| Vægt (metrisk) | 24 g |
| Linse materiale | Polycarbonate |
| Vis mere |