Få mere at vide
SJG-136, MedChemExpress
Artikelnummer. 16498697
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16498697 5 mg
16493426 25 mg
16416108 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16498697 Leverandør Medchem Express Leverandørnr. HY145735mg
MedChemExpress SJG-136 is a DNA cross-linking agent, with an XL50 of 45 nM for pBR322 DNA. SJG-136 has potent antitumor activity.
SJG-136 is a DNA cross-linking agent, with an XL50 of 45 nM for pBR322 DNA. SJG-136 has potent antitumor activity.
Tekniske data
Kemisk navn eller materiale SJG-136
CAS 232931-57-6
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C31H32N4O6
Mængde 5 mg
Synonym NSC-694501
Opløselighedsinformation DMSO : 33.33 mg/mL (59.88 mM; Need ultrasonic)
SMIL O=C1N2[C@@H](CC(C2)=C)C=NC3=CC(OCCCOC4=C(OC)C=C(C(N(CC(C5)=C)[C@@H]5C=N6)=O)C6=C4)=C(OC)C=C31
Molekylvægt (g/mol) 556.61
Formel vægt 556.61
Procent renhed 95.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease