missing translation for 'onlineSavingsMsg'
Learn More
Sitafloxacin sesquihydrate, Thermo Scientific Chemicals Product Code.: 16534414
compliance-icons

Sitafloxacin sesquihydrate, Thermo Scientific Chemicals

Product Code. p-7214492 Shop All Thermo Scientific Chemicals Products
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 16534414

Brand: Thermo Scientific Acros 464372500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163253-35-8
Molecular Formula C38H42Cl2F4N6O9
Molecular Weight (g/mol) 873.68
InChI Key MPORYQCGWFQFLA-UHFFFAOYNA-N
IUPAC Name bis(7-{7-amino-5-azaspiro[2.4]heptan-5-yl}-8-chloro-6-fluoro-1-(2-fluorocyclopropyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid) trihydrate
SMILES O.O.O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 436.84
Chemical Name or Material Sitafloxacin sesquihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0411785 Certificate TypeCertificate of Analysis Date18/12/2023 Product Code464372500,464370010
Lot NumberA0444581 Certificate TypeCertificate of Analysis Date25/08/2023 Product Code464372500,464370010
Lot NumberA0441880 Certificate TypeCertificate of Analysis Date30/05/2022 Product Code464372500,464370010
compliance-icons
Product Identifier
  • Sitafloxacin sesquihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sitafloxacin hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sitafloxacin sesquihydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.