Learn More
Sitafloxacin hydrate, MedChemExpress
MedChemExpress Sitafloxacin (DU6859a) hydrate is a potent, orally active fluoroquinolone antibiotic with in vitro activity against a broad range of gram-positive and gram-negative bacteria, including anaerobic bacteria, as well as against atypical pathogens. Sitafloxacin can be used for the research of respiratory tract infection and urinary tract infection.

725.13 DKK - 3323.51 DKK
Specifications
Chemical Name or Material | Sitafloxacin hydrate |
---|---|
Physical Form | Solid |
CAS | 163253-35-8 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16450908
![]() |
Medchem Express
HY-B0395C |
5 mg |
725.13 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16424357
![]() |
Medchem Express
HY-B0395C |
50 mg |
3323.51 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16408478
![]() |
Medchem Express
HY-B0395C |
10 mg |
1168.26 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sitafloxacin (DU6859a) hydrate is a potent, orally active fluoroquinolone antibiotic with in vitro activity against a broad range of gram-positive and gram-negative bacteria, including anaerobic bacteria, as well as against atypical pathogens. Sitafloxacin can be used for the research of respiratory tract infection and urinary tract infection.Specifications
Sitafloxacin hydrate | |
163253-35-8 | |
Research | |
COVID-19-immunoregulation | |
DMSO : 8.9 mg/mL (20.37 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (2.29 mM; ultrasonic and warming and heat to 80°C) | |
436.84 | |
98.0% |
Solid | |
White | |
C19H18ClF2N3O3.3/2H2O | |
DU6859a hydrate | |
N[C@H]1C2(CC2)CN(C(C(Cl)=C3N([C@H]4[C@@H](F)C4)C=C(C(O)=O)C5=O)=C(C=C35)F)C1.[1.5 H2O] | |
436.84 | |
Research |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.