Learn More
Tocris Bioscience™ SirReal 2
Selective inhibitor of SIRT2
Brand: Tocris Bioscience™ 5457/10
Description
SirReal 2 is a selective SIRT2 inhibitor (IC50 = 140 nM). Has minimal effect on SIRT1 and SIRT3-6. Increases α-tubulin acetylation levels in HeLa cells.Specifications
>98% | |
420.55 | |
10 mg |
SirReal 2 | |
SirReal 2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only