Learn More
Tocris Bioscience™ SirReal 2
Selective inhibitor of SIRT2
Brand:  Tocris Bioscience™ 5457/10
Encompass
Product Code. 15706856
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
SirReal 2 is a selective SIRT2 inhibitor (IC50 = 140 nM). Has minimal effect on SIRT1 and SIRT3-6. Increases α-tubulin acetylation levels in HeLa cells.
Specifications
Specifications
>98%
420.55
10 mg
SirReal 2
SirReal 2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ SirReal 2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only