missing translation for 'onlineSavingsMsg'
Learn More
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™ Product Code.: 11438553

Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™

Product Code. 11438553 Shop All Fisher Chemical Products
25 L, Plastic container, 25 L
Click to view available options
Quantity:
2.5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
10 L
2.5 L
25 L
This item is not returnable. View return policy

Product Code. 11438553

Brand: Fisher Chemical J/7330/25

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

AgNO3, CAS Number-7761-88-8, silver nitrate, lunar caustic, argerol, silver mononitrate, silvernitrate, silver i nitrate, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, 2.5L, CHEBI:32130, Colorless, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate, 3414, 169.872

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerolShow MoreShow Less
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Packaging Plastic container
Quantity 25 L
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number2544306 Certificate TypeCertificate of Analysis Date10/07/2025 Product CodeJ/7330/24,J/7330C/90,J/7330/17,J/7330/25,J/7330/15,J/7330C/05
Lot Number2542600 Certificate TypeCertificate of Analysis Date25/06/2025 Product CodeJ/7330/24,J/7330C/90,J/7330/17,J/7330/25,J/7330/15,J/7330C/05
Lot Number2538869 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/7330/24,J/7330/17,J/7330C/90,J/7330/25,J/7330/15,J/7330C/05
Lot Number2535738 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/7330/24,J/7330/17,J/7330C/90,J/7330/25,J/7330/15,J/7330C/05
Lot Number2533455 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/7330/24,J/7330/17,J/7330C/90,J/7330/25,J/7330/15,J/7330C/05
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.