Learn More
Macherey-Nagel™ Valco One-Piece Fused Silica Adapters
Recommended for use in fittings where the polyimide ferrule need not be removed. Macherey-Nagel™ Valco One-Piece Fused Silica Adapters are used in capillary gas chromatography (GC) and can be connected using a compatible fitting. Available in two sizes for a range of capillary tube diameters
Brand: Macherey-Nagel™ 724244
Additional Details : Weight : 0.01000kg
Description
- Two sizes: 1/32 or 1/16 in.
- Each size available for a variety of capillary outer diameters
- Supplied in packs of five (5)
Specifications
Adapter | |
1/32 in. | |
FS0.5-5 | |
Capillary GC of 0.4 to 0.5 mm O.D. |
Black | |
Capillary Gas Chromatography | |
5 Pack | |
Silica |