Learn More
Macherey-Nagel™ Valco One-Piece Fused Silica Adapters
Recommended for use in fittings where the polyimide ferrule need not be removed. Macherey-Nagel™ Valco One-Piece Fused Silica Adapters are used in capillary gas chromatography (GC) and can be connected using a compatible fitting. Available in two sizes for a range of capillary tube diameters
Brand:  Macherey-Nagel™ 724408
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10462371
  • 995.00 DKK / Pack of 5
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • Two sizes: 1/32 or 1/16 in.
  • Each size available for a variety of capillary outer diameters
  • Supplied in packs of five (5)
Specifications
Specifications
Adapter
1/16 in.
FS1.8-5
Tubing 0.8 mm O.D.
Black
Capillary Gas Chromatography
5 Pack
Silica