Learn More
Invitrogen™ Silencer™ Non-Standard Pre-Designed siRNA
Product Code. 15551154
Change view
Click to view available options
Quantity:
custom scale
Unit Size:
Each
Product Code. Quantity Unit Size
15551154 custom scale Each
1 options
Product Code. 15551154 Supplier Invitrogen™ Supplier No. AM16999
 Restricted Item
Forventet forsendelse: 11-12-2026
Classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
  • Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
  • Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
RNAi Type siRNA
Product Type siRNA
Quantity custom scale