missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ SI-2 hydrochloride, >98%

Beskrivelse
Potent steroid receptor coactivator 3 (SRC-3) inhibitor. Reduces SRC1, SRC2 and SRC3 levels in breast cancer cell lines. Inhibits cell growth (IC50 = 3.4 nM), induces apoptosis and attenuates migration of MDA-MB48 breast cancer cells in vitro. Inhibits tumor growth in a MDA-MB48 cell breast cancer mouse model. Orally available.
Specifications
Specifications
| Kemisk navn eller materiale | (3S)-4-[[4-(2-Butyn-1-yloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-3-thiomorpholinecarboxamide |
| Mængde | 10 mg |
| Opløselighed | Soluble to 50mM in water and to 20mM in DMSO |
| Formel vægt | 301.78 |
| Renhed | >98% |
| Molekylvægt (g/mol) | 398.492 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?